This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF639 SIRNA Assays ›
Gene Symbol:
ZNF639
Gene Name:
zinc finger protein 639
Gene Aliases:
ANC-2H01, ANC_2H01, ZASC1
Chromosome Location:
Chr.3: 179322910 - 179335535 on Build GRCh38
UniGene ID:
Hs.632578
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 124957

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF639
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303425.1 NP_001290354.1 7 980
NM_001303426.1 NP_001290355.1 6 936
NM_016331.2 NP_057415.1 7 998
XM_017006550.1 XP_016862039.1 7 1643
XM_017006551.1 XP_016862040.1 7 1298
XM_017006552.1 XP_016862041.1 7 989
XM_017006553.1 XP_016862042.1 7 1406
GenBank mRNA AB097862.1 - 7 998
AF003924.1 - 7 998
BC020500.1 - 6 771 3912907
BC026181.1 - 6 901 4794621
DQ323613.1 - 3 407

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
positive regulation of cell growth
negative regulation by host of viral transcription
positive regulation by host of viral transcription
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
viral entry into host cell
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein self-association
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rp79v:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline