siRNA Details
| Gene Symbol: TMEM40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284406.1 | NP_001271335.1 | 4 | 464 | ||
| NM_018306.3 | NP_060776.2 | 4 | 376 | |||
| XM_011533937.2 | XP_011532239.1 | 5 | 631 | |||
| GenBank mRNA | AK092470.1 | - | 4 | 376 | ||
| AK301984.1 | - | 4 | 464 | |||

TMEM40
Genomic Map