This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CYTL1 SIRNA Assays ›
Gene Symbol:
CYTL1
Gene Name:
cytokine like 1
Gene Aliases:
C17, C4orf4
Chromosome Location:
Chr.4: 5014586 - 5019470 on Build GRCh38
UniGene ID:
Hs.13872
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125197

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CYTL1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018659.2 NP_061129.1 4 607
XM_017008299.1 XP_016863788.1 5 722
GenBank mRNA AF193766.1 - 4 610
AF274956.1 - 4 467
AY359101.1 - 4 608
BC031391.1 - 4 583 5186334

Back To Top

More Information


Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

chondrocyte differentiation
signal transduction
positive regulation of transcription from RNA polymerase II promoter
inner ear development
chondroitin sulfate proteoglycan biosynthetic process
positive regulation of sequence-specific DNA binding transcription factor activity
cartilage homeostasis
receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-9rq9x:80/100.66.78.177:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0