siRNA Details
| Gene Symbol: MAGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022149.4 | NP_071432.2 | 1 | 898 | ||
| GenBank mRNA | AF295378.1 | - | 1 | 850 | ||
| AF320910.1 | - | 1 | 769 | |||
| BC010056.2 | - | 1 | 855 | 3943384 | ||
| BQ058163.1 | - | 1 | 504 | |||

MAGEF1
Genomic Map