This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RMND5A SIRNA Assays ›
Gene Symbol:
RMND5A
Gene Name:
required for meiotic nuclear division 5 homolog A
Gene Aliases:
CTLH, GID2, GID2A, RMD5, p44CTLH
Chromosome Location:
Chr.2: 86720291 - 86778041 on Build GRCh38
UniGene ID:
Hs.75277
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125392

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RMND5A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022780.3 NP_073617.1 3 757
GenBank mRNA AK022815.1 - 3 734
BC012165.1 - 3 478 4636136
BC047668.1 - 3 687 6066798

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gskzc:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline