This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FNDC11 SIRNA Assays ›
Gene Symbol:
FNDC11
Gene Name:
fibronectin type III domain containing 11
Gene Aliases:
C20orf195
Chromosome Location:
Chr.20: 63553020 - 63556712 on Build GRCh38
UniGene ID:
Hs.197755
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125417

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FNDC11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319152.1 NP_001306081.1 2 796
NM_001319153.1 NP_001306082.1 2 798
NM_024059.3 NP_076964.1 2 814
XM_011529041.2 XP_011527343.1 2 1351
XM_017028060.1 XP_016883549.1 2 950
GenBank mRNA AK125168.1 - 1 2671
BC000912.2 - 2 814 3048411
BI828873.1 - 2 790
BM554007.1 - 2 775

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline