This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ARL14 SIRNA Assays ›
Gene Symbol:
ARL14
Gene Name:
ADP ribosylation factor like GTPase 14
Gene Aliases:
ARF7
Chromosome Location:
Chr.3: 160677160 - 160678448 on Build GRCh38
UniGene ID:
Hs.287702
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125581

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ARL14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025047.2 NP_079323.1 1 953
GenBank mRNA AK026248.1 - 1 954
BC034354.1 - 1 936 4747382

Back To Top

More Information


Panther Classification:

Molecular Function -

G-protein

Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
protein binding
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline