This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other CEP63 SIRNA Assays ›
Gene Symbol:
CEP63
Gene Name:
centrosomal protein 63
Gene Aliases:
SCKL6
Chromosome Location:
Chr.3: 134485733 - 134629607 on Build GRCh38
UniGene ID:
Hs.443301
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125610
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP63
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042383.1
NP_001035842.1
5, 6
794
NM_001042384.1
NP_001035843.1
5, 6
794
NM_001042400.1
NP_001035859.1
5, 6
622
NM_025180.3
NP_079456.2
6, 7
898
XM_005247795.4
XP_005247852.1
5, 6
571
XM_005247797.3
XP_005247854.1
5, 6
826
XM_005247799.3
XP_005247856.1
5, 6
639
XM_006713760.3
XP_006713823.1
5, 6
503
XM_011513194.2
XP_011511496.1
5, 6
620
XM_011513197.2
XP_011511499.1
4, 5
454
XM_017007244.1
XP_016862733.1
5, 6
614
XM_017007245.1
XP_016862734.1
5, 6
611
XM_017007246.1
XP_016862735.1
5, 6
503
XM_017007247.1
XP_016862736.1
5, 6
620
XM_017007248.1
XP_016862737.1
5, 6
787
XM_017007249.1
XP_016862738.1
6, 7
876
XM_017007250.1
XP_016862739.1
5, 6
612
XM_017007251.1
XP_016862740.1
5, 6
610
XM_017007252.1
XP_016862741.1
5, 6
545
XM_017007253.1
XP_016862742.1
5, 6
606
XM_017007254.1
XP_016862743.1
5, 6
503
XM_017007255.1
XP_016862744.1
5, 6
826
XM_017007256.1
XP_016862745.1
6, 7
876
XM_017007257.1
XP_016862746.1
5, 6
608
XM_017007258.1
XP_016862747.1
5, 6
503
XM_017007259.1
XP_016862748.1
5, 6
620
XM_017007260.1
XP_016862749.1
5, 6
512
XM_017007261.1
XP_016862750.1
5, 6
571
XM_017007262.1
XP_016862751.1
5, 6
620
XM_017007263.1
XP_016862752.1
5, 6
826
XM_017007264.1
XP_016862753.1
6, 7
876
XM_017007265.1
XP_016862754.1
5, 6
604
XM_017007266.1
XP_016862755.1
5, 6
620
XM_017007267.1
XP_016862756.1
5, 6
787
XM_017007268.1
XP_016862757.1
5, 6
791
XM_017007269.1
XP_016862758.1
6, 7
894
GenBank mRNA
AK023448.1
-
5, 6
568
AK023738.1
-
5, 6
735
AK056465.1
-
6, 7
876
BC014050.2
-
5, 6
783
3639744
BC068997.1
-
5, 6
542
5951988
BI561425.1
-
5, 6
625
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
DNA damage checkpoint
G2/M transition of mitotic cell cycle
mitotic nuclear division
centriole replication
signal transduction in response to DNA damage
spindle assembly
cell division
de novo centriole assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline