This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DMKN SIRNA Assays ›
Gene Symbol:
DMKN
Gene Name:
dermokine
Gene Aliases:
UNQ729, ZD52F10
Chromosome Location:
Chr.19: 35497217 - 35513678 on Build GRCh38
UniGene ID:
Hs.417795
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125798

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DMKN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126056.2 NP_001119528.2 3, 4 859
NM_001126057.2 NP_001119529.2 3, 4 859
NM_001126058.2 NP_001119530.2 3, 4 859
NM_001190347.1 NP_001177276.1 3, 4 859
NM_001190348.1 NP_001177277.1 3, 4 859
NM_001190349.1 NP_001177278.1 3, 4 859
NM_033317.4 NP_201574.3 3, 4 859
XM_011527494.2 XP_011525796.1 3, 4 879
XM_011527495.2 XP_011525797.1 3, 4 879
XM_011527496.2 XP_011525798.1 3, 4 879
XM_011527497.2 XP_011525799.1 3, 4 879
XM_011527498.2 XP_011525800.1 3, 4 879
XM_011527499.2 XP_011525801.1 3, 4 879
XM_011527500.2 XP_011525802.1 3, 4 879
XM_011527501.2 XP_011525803.1 3, 4 879
XM_011527502.2 XP_011525804.1 3, 4 879
XM_011527503.2 XP_011525805.1 3, 4 879
XM_011527504.2 XP_011525806.1 3, 4 879
XM_011527505.2 XP_011525807.1 3, 4 879
XM_011527506.2 XP_011525808.1 3, 4 879
XM_011527507.2 XP_011525809.1 3, 4 879
XM_011527508.2 XP_011525810.1 3, 4 879
XM_011527509.2 XP_011525811.1 3, 4 879
XM_011527510.2 XP_011525812.1 3, 4 879
XM_011527511.2 XP_011525813.1 3, 4 879
XM_011527512.2 XP_011525814.1 3, 4 879
GenBank mRNA AY358412.1 - 3, 4 859
AY622965.1 - 3, 4 676
AY789695.1 - 3, 4 682
AY789696.1 - 3, 4 682
AY789697.1 - 3, 4 682
BC035311.1 - 3, 4 853 4752921
EF158319.1 - 3, 4 682
EF158320.1 - 3, 4 683
EF158321.1 - 3, 4 676
EF158322.1 - 3, 4 681

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

cornified envelope assembly
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rp79v:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline