This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SHKBP1 SIRNA Assays ›
Gene Symbol:
SHKBP1
Gene Name:
SH3KBP1 binding protein 1
Gene Aliases:
PP203, Sb1
Chromosome Location:
Chr.19: 40576851 - 40591399 on Build GRCh38
UniGene ID:
Hs.26506
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 125876

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SHKBP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138392.3 NP_612401.2 5 349
XM_006723474.1 XP_006723537.1 5 349
XM_006723475.1 XP_006723538.1 5 349
XM_011527492.1 XP_011525794.1 5 349
XM_011527493.1 XP_011525795.1 4 245
XM_017027473.1 XP_016882962.1 5 349
XM_017027474.1 XP_016882963.1 4 245
GenBank mRNA AF258553.1 - 5 348
AK075057.1 - 5 345
AK123695.1 - 3 327
AK300819.1 - 5 345
AK312748.1 - 5 313
BC022855.1 - 5 320 4102901
DB038927.1 - 5 349

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

positive regulation of epidermal growth factor receptor signaling pathway
protein homooligomerization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0