This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NTN5 SIRNA Assays ›
Gene Symbol:
NTN5
Gene Name:
netrin 5
Gene Aliases:
-
Chromosome Location:
Chr.19: 48661407 - 48673119 on Build GRCh38
UniGene ID:
Hs.326217
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126043

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NTN5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145807.2 NP_665806.1 7 1674
XM_017026274.1 XP_016881763.1 6 1900
GenBank mRNA BC018654.2 - 7 1600 2961115
BC018697.2 - 7 1600 2961115
BC033207.1 - 7 1600 4903484

Back To Top

More Information


Panther Classification:

Molecular Function -

extracellular matrix protein

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-rlpqk:80/100.66.76.251:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline