siRNA Details
| Gene Symbol: WDR49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178824.3 | NP_849146.1 | 4, 5 | 797 | ||
| GenBank mRNA | AK294387.1 | - | 4, 5 | 685 | ||
| BC035512.2 | - | 4, 5 | 797 | 5199615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR49| Gene Symbol: WDR49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178824.3 | NP_849146.1 | 4, 5 | 797 | ||
| GenBank mRNA | AK294387.1 | - | 4, 5 | 685 | ||
| BC035512.2 | - | 4, 5 | 797 | 5199615 | ||