This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C3ORF67 SIRNA Assays ›
Gene Symbol:
C3orf67
Gene Name:
chromosome 3 open reading frame 67
Gene Aliases:
-
Chromosome Location:
Chr.3: 58716415 - 59050078 on Build GRCh38
UniGene ID:
Hs.368434
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 126591

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C3orf67
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198463.2 NP_940865.1 7 647
XM_005264929.2 XP_005264986.1 7 1027
XM_006713013.3 XP_006713076.1 7 1027
XM_011533453.2 XP_011531755.1 7 1027
XM_011533455.2 XP_011531757.1 7 1027
XM_011533456.1 XP_011531758.1 7 736
XM_011533457.1 XP_011531759.1 5 516
XM_011533458.2 XP_011531760.1 4 399
XM_011533459.2 XP_011531761.1 7 1027
XM_011533460.2 XP_011531762.1 2 166
XM_011533461.1 XP_011531763.1 4 359
XM_017005848.1 XP_016861337.1 3 272
XM_017005849.1 XP_016861338.1 6 870
XM_017005850.1 XP_016861339.1 6 548
XM_017005851.1 XP_016861340.1 6 873
GenBank mRNA AK124111.1 - 7 649
BC050317.1 - 7 1024
BC132815.1 - 5 461 40146837
BC151142.1 - 5 461 9020498

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline