This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C7ORF43 SIRNA Assays ›
Gene Symbol:
C7orf43
Gene Name:
chromosome 7 open reading frame 43
Gene Aliases:
-
Chromosome Location:
Chr.7: 100154420 - 100158721 on Build GRCh38
UniGene ID:
Hs.533139
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 127123

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C7orf43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303470.1 NP_001290399.1 2 194
NM_018275.4 NP_060745.3 2 694
GenBank mRNA AK054872.1 - 2 190
AK310170.1 - 3 458
BC002749.2 - 2 94
BC015722.2 - 2 652 3920412

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024969, FQ211718, BC091263.
This siRNA also targets the following transcripts in Mouse: NM_153161, BC037034, AK046882, AK138960, CD354602.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline