siRNA Details
| Gene Symbol: CXorf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024689.2 | NP_078965.2 | 3 | 822 | ||
| GenBank mRNA | AK024165.1 | - | 3 | 822 | ||
| BC035375.1 | - | 3 | 758 | |||
| BC137528.1 | - | 3 | 810 | |||
| BC137529.1 | - | 3 | 810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CXorf36| Gene Symbol: CXorf36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024689.2 | NP_078965.2 | 3 | 822 | ||
| GenBank mRNA | AK024165.1 | - | 3 | 822 | ||
| BC035375.1 | - | 3 | 758 | |||
| BC137528.1 | - | 3 | 810 | |||
| BC137529.1 | - | 3 | 810 | |||