siRNA Details
| Gene Symbol: GTPBP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042717.2 | NP_001036182.1 | 8 | 1378 | ||
| NM_033107.3 | NP_149098.2 | 10 | 1615 | |||
| GenBank mRNA | AF351613.1 | - | 8 | 1379 | ||
| AK095561.1 | - | 10 | 1605 | |||
| AK125553.1 | - | 1 | 612 | |||
| AK294325.1 | - | 8 | 1328 | |||
| BC144162.1 | - | 8 | 1324 | |||
| CR933597.1 | - | 9 | 1429 | |||

GTPBP10
Genomic Map