siRNA Details
| Gene Symbol: FAM104B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166699.1 | NP_001160171.1 | 3, 4 | 299 | ||
| NM_138362.3 | NP_612371.2 | 3, 4 | 296 | |||
| NR_030722.1 | 3, 4 | 303 | ||||
| XM_017029953.1 | XP_016885442.1 | 3, 4 | 393 | |||
| GenBank mRNA | AK000441.1 | - | 3, 4 | 280 | ||
| BC000919.2 | - | 3, 4 | 274 | 3449838 | ||
| BC017571.1 | - | 3, 4 | 284 | 4105917 | ||
| CK004182.1 | - | 3, 4 | 293 | |||

FAM104B
Genomic Map