This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SSMEM1 SIRNA Assays ›
Gene Symbol:
SSMEM1
Gene Name:
serine rich single-pass membrane protein 1
Gene Aliases:
C7orf45
Chromosome Location:
Chr.7: 130205399 - 130216869 on Build GRCh38
UniGene ID:
Hs.351816
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 127925

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SSMEM1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145268.3 NP_660311.1 3 572
XM_005250141.3 XP_005250198.1 3 479
XM_011515794.2 XP_011514096.1 3 436
XM_011515795.2 XP_011514097.1 3 429
GenBank mRNA AK097635.1 - 3 558
BC017587.2 - 3 576 4838989

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ztldj:80/100.66.73.158:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline