This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LINGO2 SIRNA Assays ›
Gene Symbol:
LINGO2
Gene Name:
leucine rich repeat and Ig domain containing 2
Gene Aliases:
LERN3, LRRN6C
Chromosome Location:
Chr.9: 27937617 - 29213981 on Build GRCh38
UniGene ID:
Hs.444665
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 128021

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINGO2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258282.1 NP_001245211.1 7 1256
NM_152570.2 NP_689783.1 7 1336
XM_011517719.1 XP_011516021.1 6 2204
XM_011517724.2 XP_011516026.1 5 5223
XM_011517728.2 XP_011516030.1 2 5002
XM_017014303.1 XP_016869792.1 11 5107
XM_017014304.1 XP_016869793.1 4 7627
XM_017014305.1 XP_016869794.1 5 7210
XM_017014306.1 XP_016869795.1 3 4371
XM_017014307.1 XP_016869796.1 2 1844
GenBank mRNA AK056372.1 - 7 1336
AK172834.1 - 1 336
AK290972.1 - 2 867
AY358114.1 - 2 890
BC137514.1 - 7 1196 9021514
BC144676.1 - 7 1196 9053210

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

axonogenesis
positive regulation of synapse assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline