siRNA Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181489.5 | NP_852466.1 | 8 | 2899 | ||
| XM_005265102.2 | XP_005265159.1 | 7 | 2757 | |||
| XM_005265103.3 | XP_005265160.1 | 5 | 2029 | |||
| XM_011533674.2 | XP_011531976.1 | 7 | 2722 | |||
| XM_011533675.2 | XP_011531977.1 | 5 | 1993 | |||
| XM_017006345.1 | XP_016861834.1 | 7 | 2778 | |||
| XM_017006346.1 | XP_016861835.1 | 7 | 2721 | |||
| GenBank mRNA | AY262260.1 | - | 6 | 2702 | ||
| AY295873.2 | - | 8 | 2899 | |||
| BC101486.1 | - | 6 | 2613 | 8068992 | ||
| BC143439.1 | - | 6 | 2578 | 9051947 | ||

ZNF445
Genomic Map