This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SSX2IP SIRNA Assays ›
Gene Symbol:
SSX2IP
Gene Name:
SSX family member 2 interacting protein
Gene Aliases:
ADIP
Chromosome Location:
Chr.1: 84643707 - 84690757 on Build GRCh38
UniGene ID:
Hs.22587
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 128682

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SSX2IP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166293.1 NP_001159765.1 6 890
NM_001166294.1 NP_001159766.1 5 758
NM_001166295.1 NP_001159767.1 5 886
NM_001166417.1 NP_001159889.1 7 1068
NM_014021.3 NP_054740.3 7 940
XM_005270427.1 XP_005270484.1 6 829
XM_005270428.1 XP_005270485.1 6 787
XM_005270429.1 XP_005270486.1 6 829
XM_006710331.1 XP_006710394.1 6 747
XM_011540610.1 XP_011538912.1 7 837
XM_011540612.2 XP_011538914.1 7 878
XM_017000234.1 XP_016855723.1 6 818
XM_017000235.1 XP_016855724.1 6 796
XM_017000236.1 XP_016855725.1 6 812
XM_017000237.1 XP_016855726.1 5 601
GenBank mRNA AB023140.1 - 6 878
AK001710.1 - 2 100
AK021515.1 - 4 1463
AK093173.1 - 7 825
AK292475.1 - 6 778
AK294057.1 - 5 645
AK316472.1 - 5 664
AL133046.1 - 7 880
AY367055.1 - 7 803
AY651262.1 - 4 463
BC033637.1 - 6 765 5578582
BC035580.1 - 5 699
BC064389.1 - 6 854 5745142

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
regulation of Rac protein signal transduction
intraciliary transport involved in cilium morphogenesis
cilium assembly
centrosome organization
regulation of cell motility
protein binding
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0