This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GLIPR2 SIRNA Assays ›
Gene Symbol:
GLIPR2
Gene Name:
GLI pathogenesis related 2
Gene Aliases:
C9orf19, GAPR-1, GAPR1
Chromosome Location:
Chr.9: 36136536 - 36163913 on Build GRCh38
UniGene ID:
Hs.493819
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 128754

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GLIPR2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287010.1 NP_001273939.1 4 1958
NM_001287011.1 NP_001273940.1 5 2166
NM_001287012.1 NP_001273941.1 3 1854
NM_001287013.1 NP_001273942.1 5 1873
NM_001287014.1 NP_001273943.1 2 1582
NM_022343.3 NP_071738.1 5 2036
NR_104637.1 7 2417
NR_104638.1 5 1828
NR_104639.1 6 2171
NR_104640.1 5 2058
NR_104641.1 2 1745
GenBank mRNA AK074488.1 - 5 1442
AK075091.1 - 5 1837
AK091288.1 - 6 2650
AY039756.1 - 5 1827
BC017918.1 - 5 1260 4296721
CR749781.1 - 7 2218

Back To Top

More Information


Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of epithelial cell migration
positive regulation of epithelial to mesenchymal transition
positive regulation of ERK1 and ERK2 cascade
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-r4kfh:80/100.66.79.35:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0