siRNA Details
| Gene Symbol: FAM219A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184940.1 | NP_001171869.1 | 6 | 993 | ||
| NM_001184941.1 | NP_001171870.1 | 6 | 990 | |||
| NM_001184942.1 | NP_001171871.1 | 6 | 957 | |||
| NM_001184943.1 | NP_001171872.1 | 6 | 939 | |||
| NM_001184945.1 | NP_001171874.1 | 6 | 906 | |||
| NM_147202.1 | NP_671735.1 | 6 | 942 | |||
| XM_005251402.4 | XP_005251459.1 | 7 | 1200 | |||
| XM_011517797.2 | XP_011516099.1 | 6 | 1000 | |||
| XM_011517798.2 | XP_011516100.1 | 6 | 949 | |||
| XM_011517799.2 | XP_011516101.1 | 6 | 798 | |||
| GenBank mRNA | AK091262.1 | - | 2 | 333 | ||
| AK096350.1 | - | 6 | 942 | |||
| AK123887.1 | - | 7 | 804 | |||
| AK294054.1 | - | 6 | 957 | |||
| AK299882.1 | - | 6 | 989 | |||
| BC025348.1 | - | 2 | 350 | |||
| BC041009.2 | - | 6 | 906 | 5284905 | ||

FAM219A
Genomic Map