This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF440 SIRNA Assays ›
Gene Symbol:
ZNF440
Gene Name:
zinc finger protein 440
Gene Aliases:
-
Chromosome Location:
Chr.19: 11814241 - 11835617 on Build GRCh38
UniGene ID:
Hs.418192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 129440

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF440
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152357.2 NP_689570.2 4 2939
XM_005259731.3 XP_005259788.1 4 2999
XM_017026253.1 XP_016881742.1 3 3324
XM_017026254.1 XP_016881743.1 3 2817
GenBank mRNA AK092559.1 - 1 1035
AK095252.1 - 4 2939
BC016371.1 - 1 1036
BC035760.1 - 4 2922 5556302

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline