siRNA Details
| Gene Symbol: BEND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318539.1 | NP_001305468.1 | 3 | 795 | ||
| NM_152731.2 | NP_689944.2 | 3 | 795 | |||
| XM_011514346.2 | XP_011512648.1 | 3 | 783 | |||
| XM_017010406.1 | XP_016865895.1 | 3 | 346 | |||
| XM_017010407.1 | XP_016865896.1 | 3 | 329 | |||
| XM_017010408.1 | XP_016865897.1 | 4 | 876 | |||
| GenBank mRNA | AK054724.1 | - | 3 | 480 | ||
| AK303705.1 | - | 2 | 165 | |||
| BC036119.1 | - | 3 | 644 | |||
| DC411761.1 | - | 2 | 165 | |||

BEND6
Genomic Map