siRNA Details
| Gene Symbol: NLRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176820.3 | NP_789790.2 | 2 | 1127 | ||
| GenBank mRNA | AY154464.1 | - | 2 | 1099 | ||
| BC136583.1 | - | 2 | 1119 | 9020573 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NLRP9| Gene Symbol: NLRP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176820.3 | NP_789790.2 | 2 | 1127 | ||
| GenBank mRNA | AY154464.1 | - | 2 | 1099 | ||
| BC136583.1 | - | 2 | 1119 | 9020573 | ||