This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MANEA SIRNA Assays ›
Gene Symbol:
MANEA
Gene Name:
mannosidase endo-alpha
Gene Aliases:
ENDO, hEndo
Chromosome Location:
Chr.6: 95577497 - 95609452 on Build GRCh38
UniGene ID:
Hs.734632
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 130650

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MANEA
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024641.3 NP_078917.2 5 1485
XM_005267147.3 XP_005267204.1 5 1878
GenBank mRNA AJ577574.1 - 4 1350
AK022900.1 - 4 991
AK291940.1 - 5 1416
AY372528.1 - 4 1305
BC137014.1 - 4 1354 9021007
BC137016.1 - 4 1354 9021009
BC146671.1 - 5 1420 40148201
BC150199.1 - 5 1420 40148811
BX537398.1 - 5 1416
BX640869.1 - 5 1446
CR936827.1 - 5 1420

Back To Top

More Information


Gene Ontology Categories:

Process(es)

glycoprotein endo-alpha-1,2-mannosidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-mfvw6:80/100.66.76.149:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0