siRNA Details
| Gene Symbol: UBXN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171091.1 | NP_001164562.1 | 6, 7 | 621 | ||
| NM_025241.2 | NP_079517.1 | 6, 7 | 723 | |||
| XM_017027325.1 | XP_016882814.1 | 7, 8 | 1154 | |||
| GenBank mRNA | AF272893.1 | - | 6, 7 | 722 | ||
| AF272894.1 | - | 6, 7 | 612 | |||
| AK090473.1 | - | 1 | 2696 | |||
| AL833695.1 | - | 1, 2 | 578 | |||
| BC007414.2 | - | 2, 3 | 183 | |||
| BC008288.1 | - | 6, 7 | 621 | 3659797 | ||

UBXN6
Genomic Map