This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other THAP7 SIRNA Assays ›
Gene Symbol:
THAP7
Gene Name:
THAP domain containing 7
Gene Aliases:
-
Chromosome Location:
Chr.22: 20999772 - 21002115 on Build GRCh38
UniGene ID:
Hs.512756
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 130916

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: THAP7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008695.1 NP_001008695.1 5 1039
NM_030573.2 NP_085050.2 4 1213
GenBank mRNA AK024609.1 - 2 1780
AK025906.1 - 2 1545
AK300715.1 - 5 828
BC004346.1 - 4 1184 3633743
CR456343.1 - 4 1031

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
negative regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding
protein N-terminus binding
C2H2 zinc finger domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0