This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TULP1 SIRNA Assays ›
Gene Symbol:
TULP1
Gene Name:
tubby like protein 1
Gene Aliases:
LCA15, RP14, TUBL1
Chromosome Location:
Chr.6: 35497874 - 35512902 on Build GRCh38
UniGene ID:
Hs.485208
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 13163

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TULP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289395.1 NP_001276324.1 7, 8 701
NM_003322.4 NP_003313.3 8, 9 860
GenBank mRNA AB593123.1 - 8, 9 860
AB593124.1 - 8, 9 846
BC032714.1 - 7, 8 686 5495136
BC065261.1 - 7, 8 669 6144003
CN355966.1 - 3, 4 483
U82468.1 - 8, 9 852

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
phagocytosis, recognition
visual perception
dendrite development
eye photoreceptor cell development
photoreceptor cell maintenance
positive regulation of phagocytosis
detection of light stimulus involved in visual perception
retina development in camera-type eye
receptor localization to nonmotile primary cilium
protein localization to photoreceptor outer segment
protein binding
phosphatidylinositol-4,5-bisphosphate binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-brvb8:80/100.66.78.89:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0