siRNA Details
| Gene Symbol: LRSAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005373.3 | NP_001005373.1 | 19, 20 | 1798 | ||
| NM_001005374.3 | NP_001005374.1 | 18, 19 | 1503 | |||
| NM_138361.5 | NP_612370.3 | 18, 19 | 2057 | |||
| XM_006717316.3 | XP_006717379.1 | 19, 20 | 1798 | |||
| XM_017015283.1 | XP_016870772.1 | 19, 20 | 2797 | |||
| XM_017015284.1 | XP_016870773.1 | 19, 20 | 1793 | |||
| GenBank mRNA | AK056305.1 | - | 7, 8 | 648 | ||
| AK091589.1 | - | 18, 19 | 1805 | |||
| AK226106.1 | - | 19, 20 | 1784 | |||
| AY358830.1 | - | 19, 20 | 1763 | |||
| BC009239.2 | - | 18, 19 | 1503 | 2988664 | ||

LRSAM1
Genomic Map