This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C21ORF91 SIRNA Assays ›
Gene Symbol:
C21orf91
Gene Name:
chromosome 21 open reading frame 91
Gene Aliases:
C21orf14, C21orf38, CSSG1, EURL, YG81
Chromosome Location:
Chr.21: 17788967 - 17819386 on Build GRCh38
UniGene ID:
Hs.293811
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 131939

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C21orf91
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100420.1 NP_001093890.1 5 961
NM_001100421.1 NP_001093891.1 4 898
NM_017447.3 NP_059143.3 5 958
GenBank mRNA AF239726.1 - 4 851
AK023825.1 - 5 926
AK225123.1 - 4 868
BC015468.2 - 4 874 3916462

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hr4r9:80/100.66.79.48:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0