This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF407 SIRNA Assays ›
Gene Symbol:
ZNF407
Gene Name:
zinc finger protein 407
Gene Aliases:
-
Chromosome Location:
Chr.18: 74597735 - 75065672 on Build GRCh38
UniGene ID:
Hs.536490
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 132068

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF407
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146189.1 NP_001139661.1 1 2942
NM_001146190.1 NP_001139662.1 1 2942
NM_017757.2 NP_060227.2 1 2942
XM_005266726.4 XP_005266783.1 2 3141
XM_006722500.3 XP_006722563.1 2 3006
XM_011526068.2 XP_011524370.1 2 6648
XM_011526069.2 XP_011524371.1 2 3006
XM_011526070.1 XP_011524372.1 2 3006
XM_017025838.1 XP_016881327.1 2 3006
GenBank mRNA AK000314.1 - 1 567

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline