This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRRN3 SIRNA Assays ›
Gene Symbol:
LRRN3
Gene Name:
leucine rich repeat neuronal 3
Gene Aliases:
FIGLER5, NLRR-3, NLRR3
Chromosome Location:
Chr.7: 111091006 - 111125454 on Build GRCh38
UniGene ID:
Hs.3781
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 132450

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099658.1 NP_001093128.1 3 3085
NM_001099660.1 NP_001093130.1 4 3192
NM_018334.4 NP_060804.3 2 3003
GenBank mRNA AB060967.1 - 1 2452
AF134481.1 - 1 2483
AK292556.1 - 2 2592
AL442092.1 - 1 2408
AY358280.1 - 3 2625
BC035133.2 - 2 2882 5263900

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-kdjgd:80/100.66.79.206:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline