This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEMGN SIRNA Assays ›
Gene Symbol:
HEMGN
Gene Name:
hemogen
Gene Aliases:
CT155, EDAG, EDAG-1, NDR
Chromosome Location:
Chr.9: 97926791 - 97944915 on Build GRCh38
UniGene ID:
Hs.176626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 132534

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HEMGN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018437.4 NP_060907.2 4 549
NM_197978.2 NP_932095.1 3 603
XM_005252086.1 XP_005252143.1 4 511
XM_011518845.2 XP_011517147.1 4 423
XM_011518846.2 XP_011517148.1 4 533
GenBank mRNA AF130060.2 - 3 336
AF228713.2 - 4 456
AF306682.1 - 3 385
AF306683.1 - 3 351
AF322875.1 - 4 455
AK292441.1 - 4 475
AL833094.1 - 3 395
AY244805.1 - 3 549
AY255672.1 - 3 454
BC048324.1 - 4 458 5166612
BG571634.1 - 3 427
BG717036.1 - 4 494

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
regulation of osteoblast differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-wrbwd:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline