This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C12ORF4 SIRNA Assays ›
Gene Symbol:
C12orf4
Gene Name:
chromosome 12 open reading frame 4
Gene Aliases:
-
Chromosome Location:
Chr.12: 4487730 - 4538508 on Build GRCh38
UniGene ID:
Hs.302977
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 133085

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C12orf4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304811.1 NP_001291740.1 3, 4 438
NM_020374.3 NP_065107.1 3, 4 446
XM_005253716.1 XP_005253773.1 3, 4 446
XM_006718992.3 XP_006719055.1 3, 4 446
GenBank mRNA AJ272205.1 - 3, 4 389
AK074479.1 - 3, 4 446
AL834377.1 - 3, 4 409
BC016908.1 - 3, 4 389 4402263
BX641128.1 - 3, 4 439
DA195770.1 - 3, 4 411

Back To Top

More Information


Gene Ontology Categories:

Function(s)

regulation of mast cell degranulation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline