siRNA Details
| Gene Symbol: C12orf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304811.1 | NP_001291740.1 | 7 | 900 | ||
| NM_020374.3 | NP_065107.1 | 7 | 908 | |||
| XM_005253716.1 | XP_005253773.1 | 7 | 908 | |||
| XM_006718992.3 | XP_006719055.1 | 7 | 908 | |||
| GenBank mRNA | AJ272205.1 | - | 7 | 851 | ||
| AK074479.1 | - | 7 | 908 | |||
| AL834377.1 | - | 7 | 871 | |||
| BC016908.1 | - | 7 | 851 | 4402263 | ||
| BX641128.1 | - | 7 | 900 | |||

C12orf4
Genomic Map