This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRICKLE4 SIRNA Assays ›
Gene Symbol:
PRICKLE4 TOMM6
Gene Name:
prickle planar cell polarity protein 4
translocase of outer mitochondrial membrane 6
Gene Aliases:
C6orf49, OBTP, OEBT, TOMM6
OBTP, TOM6
Chromosome Location:
Chr.6: 41780762 - 41787372 on Build GRCh38
Chromosome Location:
Chr.6: 41787443 - 41789896 on Build GRCh38
UniGene ID:
Hs.715653
Hs.731633
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 134117

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRICKLE4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013397.5 NP_037529.3 8 1074
GenBank mRNA BC110458.1 - 6 1521
BC110459.1 - 6 1526 40024767
Gene Symbol: TOMM6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ420506.1 - 1 67

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline