This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RBMS3 SIRNA Assays ›
Gene Symbol:
RBMS3
Gene Name:
RNA binding motif single stranded interacting protein 3
Gene Aliases:
-
Chromosome Location:
Chr.3: 29280860 - 30010395 on Build GRCh38
UniGene ID:
Hs.221436
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 134391

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RBMS3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003792.2 NP_001003792.1 9 1196
NM_001003793.2 NP_001003793.1 9 1199
NM_001177711.1 NP_001171182.1 9 1199
NM_001177712.1 NP_001171183.1 10 1238
NM_014483.3 NP_055298.2 9 1199
XM_005265060.2 XP_005265117.1 9 1648
XM_005265061.2 XP_005265118.1 9 1651
XM_005265062.1 XP_005265119.1 9 1651
XM_005265063.1 XP_005265120.1 6 856
XM_005265065.4 XP_005265122.1 6 549
XM_017006178.1 XP_016861667.1 9 1648
XM_017006179.1 XP_016861668.1 9 1648
XM_017006180.1 XP_016861669.1 9 1651
XM_017006181.1 XP_016861670.1 9 1648
XM_017006182.1 XP_016861671.1 10 1727
GenBank mRNA AF023259.1 - 9 976
AK292789.1 - 9 875
AY236871.1 - 9 1059
AY236872.1 - 9 1056
BC117315.1 - 10 935 40125866
BC143521.1 - 10 935 9052030

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of translation
nucleotide binding
mRNA 3'-UTR binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-npfdm:80/100.66.76.105:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0