This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KIF1BP SIRNA Assays ›
Gene Symbol:
KIF1BP
Gene Name:
KIF1 binding protein
Gene Aliases:
KBP, KIAA1279, TTC20
Chromosome Location:
Chr.10: 68988721 - 69016983 on Build GRCh38
UniGene ID:
Hs.279580
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 134606

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KIF1BP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015634.3 NP_056449.1 4 750
GenBank mRNA AB033105.1 - 4 673
AK291343.1 - 4 655
BC006427.1 - 4 673
BC012180.1 - 4 647 4550085

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
nervous system development
cell differentiation
protein binding
kinesin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline