siRNA Details
| Gene Symbol: ING4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127582.1 | NP_001121054.1 | 8 | 990 | ||
| NM_001127583.1 | NP_001121055.1 | 8 | 981 | |||
| NM_001127584.1 | NP_001121056.1 | 8 | 978 | |||
| NM_001127585.1 | NP_001121057.1 | 7 | 918 | |||
| NM_001127586.1 | NP_001121058.1 | 7 | 842 | |||
| NM_016162.3 | NP_057246.2 | 8 | 987 | |||
| XM_005253698.3 | XP_005253755.1 | 9 | 1238 | |||
| XM_011520964.2 | XP_011519266.1 | 7 | 916 | |||
| XM_011520965.2 | XP_011519267.1 | 8 | 937 | |||
| XM_017019392.1 | XP_016874881.1 | 9 | 1235 | |||
| XM_017019393.1 | XP_016874882.1 | 9 | 1226 | |||
| GenBank mRNA | AF063594.1 | - | 8 | 962 | ||
| AF110645.1 | - | 8 | 966 | |||
| AF156552.1 | - | 8 | 966 | |||
| AK293169.1 | - | 7 | 903 | |||
| BC007781.2 | - | 8 | 952 | 4309278 | ||
| BC013038.2 | - | 8 | 932 | |||
| BC095434.1 | - | 8 | 951 | 30365729 | ||

ING4
Genomic Map