This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HSD17B11 SIRNA Assays ›
Gene Symbol:
HSD17B11
Gene Name:
hydroxysteroid 17-beta dehydrogenase 11
Gene Aliases:
17-BETA-HSD11, 17-BETA-HSDXI, 17BHSD11, DHRS8, PAN1B, RETSDR2, SDR16C2
Chromosome Location:
Chr.4: 87336522 - 87391303 on Build GRCh38
UniGene ID:
Hs.594923
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 134920

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HSD17B11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016245.4 NP_057329.3 2 529
XM_011532021.1 XP_011530323.1 2 478
GenBank mRNA AF126780.1 - 2 484
AF273056.1 - 2 382
AK075348.1 - 2 413
AY358553.1 - 2 396
BC008650.1 - 2 419 3854750
BC014327.1 - 2 395 3961721
BC016367.1 - 2 426 4133318
BC021673.1 - 2 400 3827935
BC036001.1 - 2 420 4715042
DA265697.1 - 2 528

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
estrogen biosynthetic process
androgen catabolic process
oxidation-reduction process
estradiol 17-beta-dehydrogenase activity
steroid dehydrogenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0