This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM53C SIRNA Assays ›
Gene Symbol:
FAM53C
Gene Name:
family with sequence similarity 53 member C
Gene Aliases:
C5orf6
Chromosome Location:
Chr.5: 138337535 - 138349729 on Build GRCh38
UniGene ID:
Hs.54056
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 135121

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM53C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135647.1 NP_001129119.1 4 898
NM_016605.2 NP_057689.1 4 873
GenBank mRNA AF251040.1 - 4 609
AK315567.1 - 4 622
BC000259.1 - 4 615 3357346
BC052993.1 - 4 838 5164799

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline