siRNA Details
| Gene Symbol: IL17D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138284.1 | NP_612141.1 | 3 | 1587 | ||
| XM_005266421.3 | XP_005266478.1 | 3 | 2031 | |||
| XM_005266422.2 | XP_005266479.1 | 2 | 1748 | |||
| GenBank mRNA | AK123964.1 | - | 3 | 1670 | ||
| AY078238.1 | - | 3 | 1587 | |||
| AY359113.1 | - | 2 | 1479 | |||
| BC036243.1 | - | 3 | 1590 | 5301608 | ||

IL17D
Genomic Map