This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXW8 SIRNA Assays ›
Gene Symbol:
FBXW8
Gene Name:
F-box and WD repeat domain containing 8
Gene Aliases:
FBW6, FBW8, FBX29, FBXO29, FBXW6
Chromosome Location:
Chr.12: 116910949 - 117032498 on Build GRCh38
UniGene ID:
Hs.624537
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 135429

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXW8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012174.1 NP_036306.1 4 510
NM_153348.2 NP_699179.2 4 708
XM_017019175.1 XP_016874664.1 4 485
XM_017019176.1 XP_016874665.1 5 717
XM_017019177.1 XP_016874666.1 5 717
GenBank mRNA AF176707.1 - 4 488
AK289621.1 - 4 501
BC037296.2 - 4 708 4821240

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Golgi organization
cell proliferation
protein ubiquitination
positive regulation of dendrite morphogenesis
spongiotrophoblast layer development
labyrinthine layer blood vessel development
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline