This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BLCAP SIRNA Assays ›
Gene Symbol:
BLCAP
Gene Name:
bladder cancer associated protein
Gene Aliases:
BC10
Chromosome Location:
Chr.20: 37517417 - 37527931 on Build GRCh38
UniGene ID:
Hs.472651
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 136024

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BLCAP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167820.1 NP_001161292.1 3 574
NM_001167821.1 NP_001161293.1 2 404
NM_001167822.2 NP_001161294.1 2 461
NM_001167823.1 NP_001161295.1 2 477
NM_001317074.1 NP_001304003.1 2 454
NM_001317075.1 NP_001304004.1 2 370
NM_006698.3 NP_006689.1 2 441
XM_017027599.1 XP_016883088.1 2 1354
GenBank mRNA AF053470.1 - 2 380
AK074498.1 - 4 471
AK124822.1 - 3 299
AL049288.1 - 2 387
BC047692.1 - 2 372 6059195
BC071704.1 - 2 366 5190373
BP293276.1 - 2 477
CB155706.1 - 2 411
DA198124.1 - 2 404
DA262965.1 - 2 441
DA283758.1 - 2 370
DA632207.1 - 3 523
DA770824.1 - 2 441
DA943717.1 - 2 410

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell cycle
apoptotic nuclear changes
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0