This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TDRKH SIRNA Assays ›
Gene Symbol:
TDRKH
Gene Name:
tudor and KH domain containing
Gene Aliases:
TDRD2
Chromosome Location:
Chr.1: 151769354 - 151790576 on Build GRCh38
UniGene ID:
Hs.144439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 136140

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TDRKH
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083963.1 NP_001077432.1 6 868
NM_001083964.1 NP_001077433.1 6 758
NM_001083965.1 NP_001077434.1 6 893
NM_006862.3 NP_006853.2 6 893
XM_005244856.4 XP_005244913.1 6 659
XM_017000122.1 XP_016855611.1 6 935
XM_017000123.1 XP_016855612.1 6 935
XM_017000124.1 XP_016855613.1 6 834
XM_017000125.1 XP_016855614.1 6 863
XM_017000126.1 XP_016855615.1 6 834
XM_017000127.1 XP_016855616.1 6 863
XM_017000128.1 XP_016855617.1 6 788
XM_017000129.1 XP_016855618.1 6 753
XM_017000130.1 XP_016855619.1 6 727
GenBank mRNA AF119121.2 - 6 881
AF227192.1 - 6 881
AK056402.1 - 6 816
AK225160.1 - 6 747
AK226092.1 - 6 806
AK295948.1 - 6 664
AK303063.1 - 6 789
AK316279.1 - 6 744
BC032690.1 - 6 800

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

male meiosis
spermatogenesis
fertilization
cell differentiation
gene silencing by RNA
piRNA metabolic process
DNA methylation involved in gamete generation
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-6qxl5:80/100.66.77.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline