This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HHLA3 SIRNA Assays ›
Gene Symbol:
HHLA3
Gene Name:
HERV-H LTR-associating 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 70353840 - 70368023 on Build GRCh38
UniGene ID:
Hs.142245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 136237

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HHLA3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031693.2 NP_001026863.1 2 353
NM_001036645.1 NP_001031722.1 2 450
NM_001036646.1 NP_001031723.1 2 349
NR_027404.1 2 349
XM_011540547.2 XP_011538849.1 3 1603
XM_011540548.2 XP_011538850.1 2 248
GenBank mRNA AF126163.1 - 2 271
AF126164.1 - 2 330
BC010922.1 - 2 331 4064190
BG773961.1 - 2 450
BI868263.1 - 2 349

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline