siRNA Details
| Gene Symbol: FNBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015308.3 | NP_056123.2 | 1, 2 | 402 | ||
| GenBank mRNA | AK054636.1 | - | 1, 2 | 225 | ||
| AK123898.1 | - | 1, 2 | 206 | |||
| AL137480.1 | - | 1, 2 | 201 | |||
| BC037404.1 | - | 1, 2 | 233 | 5140963 | ||
| DB458472.1 | - | 1, 2 | 384 | |||

FNBP4
Genomic Map