This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MORC3 SIRNA Assays ›
Gene Symbol:
MORC3
Gene Name:
MORC family CW-type zinc finger 3
Gene Aliases:
NXP2, ZCW5, ZCWCC3
Chromosome Location:
Chr.21: 36320189 - 36376646 on Build GRCh38
UniGene ID:
Hs.421150
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 136913

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MORC3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320445.1 NP_001307374.1 5 680
NM_001320446.1 NP_001307375.1 7 839
NM_015358.2 NP_056173.1 6 753
XM_011529518.2 XP_011527820.1 6 973
XM_017028311.1 XP_016883800.1 6 775
XM_017028312.1 XP_016883801.1 6 696
GenBank mRNA AK292957.1 - 6 704
AK295133.1 - 5 672
BC007311.1 - 6 710
BC094779.1 - 6 710 30527767
BC132731.1 - 6 707 40146753
BC144116.1 - 7 793
D50926.2 - 6 712

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cell aging
post-embryonic development
viral process
peptidyl-serine phosphorylation
negative regulation of fibroblast proliferation
protein stabilization
maintenance of protein location in nucleus
RNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline